Hlavní dokumenty
Z803324
Lightweight die for Specac® Atlas 15T presses
size 40 mm
About This Item
UNSPSC Code:
12000000
NACRES:
SB.32
manufacturer/tradename
Specac Ltd
technique(s)
IR spectroscopy: suitable
size
40 mm
General description
The Atlas Lightweight Evacuable Pellet Dies have been specifically designed to work with the range of Atlas automatic presses up to a maximum load rating of 25 tons. The dies are also compatible for use with Specac′s 15 and 25 ton manual hydraulic presses.
They are well suited for the preparation of solid pellets for study by X-Ray Fluorescence spectroscopy. When many samples are prepared for study, the pyhsical effort required to load and remove an Atlas Lightweight Evacuable Pellet Die into a press is greatly reduced, as the dies themselves are physically smaller and lighter in weight by comparison to the conventional 32mm and 40mm pellet dies.
Legal Information
Specac is a registered trademark of Specac Ltd.
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)