Hlavní dokumenty
Z800791
Specac® Omni Cell spacers
set of assorted PTFE spacers
About This Item
UNSPSC Code:
12000000
NACRES:
SB.32
packaging
pkg of 1 ea (2 each size)
manufacturer/tradename
Specac Ltd
technique(s)
IR spectroscopy: suitable
General description
Contains 10 assorted PTFE spacers for Specac Omni Cells, 2 of each per thickness: 0.05 mm, 0.10 mm, 0.20 mm, 0.50 mm, 1.00 mm (rectangular).
Legal Information
Specac is a registered trademark of Specac Ltd.
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)