Key Documents
Z803103
Specac® Atlas 15T manual hydraulic press
About This Item
UNSPSC Code:
12000000
NACRES:
SB.32
manufacturer/tradename
Specac Ltd
technique(s)
IR spectroscopy: suitable
General description
Specac® Atlas 15T manual hydraulic press may be used for the fabrication of free-standing bio-composites by employing two bio-polyesters with different elastic moduli and reinforced with different graphene nanoplatelets. It may also be used to compress iron oxide granules to iron oxide discs to calculate the contact angle for the adsorption of heavy metals from wastewater using granulated iron oxide.
Features and Benefits
  • Polycarbonate safety guards
  • Adjustable upper bolster
  • Adjustable pressure control valve
  • Vacuum ports
  • Pressure release valve
Legal Information
Specac is a registered trademark of Specac Ltd.
accessory
Č. produktu
Popis
Stanovení ceny
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)
Lot/Batch Number
Je nám líto, ale pro tento produkt momentálně nemáme COA k dispozici online.
Navštívit knihovnu dokumentů
Effect of graphene nano-platelet morphology on the elastic modulus of soft and hard biopolymers.
Cataldi P, et al.
Carbon, 109, 331-339 (2016)
Design, production and characterisation ofgranular adsorbent material for arsenic removalfrom contaminated wastewater.
Mangwadi C, et al.
Chemical Engineering Research and Design, 110, 70-81 (2016)