Z506699
Aldrich® macro-micro KBr pellet die
About This Item
UNSPSC Code:
12000000
NACRES:
SB.32
Doporučené produkty
technique(s)
IR spectroscopy: suitable
diam.
13 mm
General description
- For producing 13mm diameter KBr pellets. Use in conjunction with a 12 ton lab press and a vacuum pump, capable of reducing pressure to 2mm, to reduce moisture in the sample. Heavy paper inserts in a variety of smaller apertures are available separately for making micropellets with the 13mm die set and also provide a useful frame for mounting and storing the pellets.
- Hardened stainless steel piston and lower section
- Black anodized aluminum body
- Optically polished anvils
- Viton® seals
- Evacuable via 1/4in. O.D. hose connection
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Viton is a registered trademark of E. I. du Pont de Nemours and Company
related product
Č. produktu
Popis
Stanovení ceny
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)