Key Documents
ZLXLP0D01
Milli-Q
Loop-Point of Delivery-Inters-T-valve
Additional loop point to make a second Point of Delivery on the water distribution loop
About This Item
UNSPSC Code:
41104214
NACRES:
JA.13
packaging
pkg of 1 unit
compatibility
for use with Milli-Q® CLX
for use with Milli-Q® HR
for use with Milli-Q® HX
General description
This accessory kit contains parts to make a second point of delivery on the water distribution loop. Normally, the distribution loop has at least one point of use. This accessory is required when creating additional points of delivery.The kit contains an interconnector "I" Hose barb 3/8" (to fit 10x16 tubings), interconnector "T" 3/8", Oeticker rings, interconnector wall mounting bracket (to wall mount the "T" interconnector), a Clamp for interconnector and a Ball valve.
Application
To add second Point of Delivery to a distribution loop.
Features and Benefits
Genuine accessory for Loop-Point of delivery - Inters-T-valve - ZLXLP0D01
Legal Information
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany