O této položce
NACRES:
JA.13
UNSPSC Code:
41104214
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohlipackaging
pkg of 1 unit
compatibility
for use with Elix® Advantage, for use with Elix® Reference, for use with Milli-Q® Direct, for use with Milli-Q® Integral
General description
The Lab Close Kit will avoid the water purification system standstill during long inactive periods. The system when used in association with the "Lab Close" operating mode, allows you to maintain a minimum operating level.
This kit intelligently optimizes the consumption of water and electricity by your system, preventing the negative effects of nonuse.
This kit intelligently optimizes the consumption of water and electricity by your system, preventing the negative effects of nonuse.
Application
Limits unnecessary water consumption, while avoiding stagnation in the water flow and storage tank.
Features and Benefits
- Minimizes water and energy usage during vacation or long inactivity periods.
- Prevents contamination of water purification system and reservoir due to water stagnation.
- Your system is up and running immediately when you come back.
Legal Information
ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
Globální číslo obchodní položky
| Skladová položka | GTIN |
|---|---|
| LABCL0SE1 | 04053252330230 |