Key Documents
Z742375
Sigma-Aldrich
CANgas dosing valve
About This Item
UNSPSC Code:
12142100
NACRES:
NA.22
Quality Level
General description
Dosing valve without regulator function. Applicable for Messer® CANgas
Other Notes
Product is currently only available to our customers in Europe
Legal Information
Messer is a registered trademark of Messer Group GmbH
regulator
Č. produktu
Popis
Stanovení ceny
Zákazníci si také prohlíželi
Slide 1 of 7
1 of 7
Air synthetic, Messer® CANgas, 99.999%
Sigma-Aldrich
769096
Air
Helium Messer® CANGas, 99.999%
Sigma-Aldrich
768995
Helium
Neon Messer® CANGas, 99.999%
Sigma-Aldrich
769029
Neon
Xenon 99.99%, Messer® CANGas
Sigma-Aldrich
769118
Xenon
Propane 99.95%, Messer® CANGas
Sigma-Aldrich
769037
Propane