Hlavní dokumenty
23884
In-Line Pressure Regulator
Ultra-High Purity version (outlet pressure 0-100 psi, 1/8 in. stainless steel fittings)
About This Item
UNSPSC Code:
41112500
compatibility
Air, H2, N2, Ar, He
General description
Install an in-line pressure regulator downstream of a gas cylinder regulator to adjust pressure closer to the point of use. 400 psig maximum input pressure. 1/8 in. Swagelok nuts and ferrules included.
Application
Air, H2, N2, Ar, He
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)