Key Documents
Z742378
Sigma-Aldrich
CANgas outer adaptor
About This Item
UNSPSC Code:
12142100
NACRES:
NA.22
Quality Level
General description
1/8′′ NPT outer adaptor for pressure can. Applicable for Messer® CANgas
Other Notes
Product is currently only available to our customers in Europe
Legal Information
Messer is a registered trademark of Messer Group GmbH
Zákazníci si také prohlíželi
Slide 1 of 5
1 of 5
Helium Messer® CANGas, 99.999%
Sigma-Aldrich
768995
Helium
Air synthetic, Messer® CANgas, 99.999%
Sigma-Aldrich
769096
Air
Propane 99.95%, Messer® CANGas
Sigma-Aldrich
769037
Propane
Ethylene Messer® CANGas, 99.95%
Sigma-Aldrich
768987
Ethylene
Xenon 99.99%, Messer® CANGas
Sigma-Aldrich
769118
Xenon