Hlavní dokumenty
Z404136
Sigma-Aldrich
IKA® VRX basic Vibrax® orbital shaker attachments
VX 2 test tube attachment, for up to 36 test tubes or centrifugal tubes with a diameter of 16 mm, 1/cs
About This Item
UNSPSC Code:
41103811
NACRES:
NB.73
packaging
pack of 1
manufacturer/tradename
IKA 0000568900
H × W × D
115 mm × 140 mm × 145 mm
General description
PC unit mixes up to 36 test tubes or centrifuge tubes with 16 mm diameter
Legal Information
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
Vibrax is a registered trademark of IKA-Werke GmbH & Co. KG
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)