Z121517
Aldrich® cold-trap with ball joint
joint: SJ 65/40
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
Doporučené produkty
joint
joint: SJ65/40
General description
Trap 7 cm I.D. × 30 cm Length, overall 75 mm O.D. × 470 mm Length, 20 mm O.D. side tubes. Includes pinch clamp.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
replacement part
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)