Vybrat velikost
| Vám/SKU | Dostupnost | Cena |
|---|---|---|
1 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 4 380,00 Kč |
O této položce
4 380,00 Kč
joint
joint: ST/NS40/50
packaging
pkg of 1 ea
manufacturer/tradename
Synthware V164050
L
250 mm , below joint
body O.D.
50 mm
General description
Legal Information
1 of 1
Tato položka | |||
|---|---|---|---|
| description joint: ST/NS 40/50, body O.D. 50 mm, L 250 mm , below joint | description joint: ST/NS 24/40, body O.D. 30 mm, L 200 mm , below joint | description joint: ST/NS 29/42, body O.D. 32 mm, L 200 mm , below joint | description joint: ST/NS 34/45, body O.D. 40 mm, L 225 mm , below joint |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
| body O.D. 50 mm | body O.D. 30 mm | body O.D. 32 mm | body O.D. 40 mm |
| manufacturer/tradename Synthware V164050 | manufacturer/tradename Synthware V163024 | manufacturer/tradename Synthware V163229 | manufacturer/tradename Synthware V164034 |
| L 250 mm , below joint | L 200 mm , below joint | L 200 mm , below joint | L 225 mm , below joint |
| joint joint: ST/NS40/50 | joint joint: ST/NS24/40 | joint joint: ST/NS29/42 | joint joint: ST/NS34/45 |
Still not finding the right product?
Explore all of our products under Synthware™ vacuum trap with serrated hose connections
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?