Key Documents
Z106534
Supelco
Septa for open top caps
for cap size 15 mm, PTFE/red rubber
About This Item
UNSPSC Code:
41122001
NACRES:
NB.11
material
PTFE/red rubber
packaging
pkg of 100
General description
For use with cut-out top caps and Wheaton vials.