Merck
Z743323
Breathe-EASY® tube membranes for microtubes, , polyurethane, precut sizes
sterile
Synonym(e):
breathable sealing membrane, cell culture, microplates, microwell plates, multi-well plates, multi-well sealing film, plate sealers, plate sealing film, sealing film, tissue culture
Größe auswählen
100 EA
CHF 48.00
CHF 48.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
100 EA
CHF 48.00
About This Item
UNSPSC-Code:
41122410
NACRES:
NB.35
CHF 48.00
Check Cart for Availability
Sterilität
sterile
Verpackung
pack of 100 ea
Hersteller/Markenname
Diversified Biotech BMTM-1000
Allgemeine Beschreibung
Breathe-EASY polymer-type membrane offers a water vapor transmission rate of 0.7 kg/ m2/24 hours.
  • Membranes are ideal for cell growth, concentrating samples (e.g. freeze drying), and gas exchange.
  • Easy microtube cap closure. Will not bind in centrifuge rotors.
  • Formatted with an easy-to-use finger tab. Pre-cut to fit a variety of popular tube sizes. Convenient scoring makes liner removal quick and easy.
  • Membranes are gamma irradiated to ensure sterility. Free of cytotoxins.
Rechtliche Hinweise
Breathe-Easy is a registered trademark of Diversified Biotech
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Kunden haben sich ebenfalls angesehen
Slide 1 of 1