Merck
Z742618
Tube-O-Array dialyzer tray for Tube-O-DIALYZER
Größe auswählen
1 EA
CHF 227.00
CHF 227.00
Voraussichtliches Versanddatum25. Mai 2025
Größe auswählen
Ansicht ändern
1 EA
CHF 227.00
About This Item
UNSPSC-Code:
41123100
NACRES:
NB.24
CHF 227.00
Voraussichtliches Versanddatum25. Mai 2025
Hersteller/Markenname
G-BioSciences 786-145A
Verpackung
1 ea of
Allgemeine Beschreibung
Tube-O-Array Dialyzer is specifically developed for dialysis-equilibration of samples prior to 2D-gel analysis or other applications. Optimize up to 12 samples at a time. It consists of an Array Dialyzer Tray for holding 12 Tube-O-DIALYZER assemblies for simple and convenient equilibration?dialysis of the samples. It is suitable for 20μl to 2.5ml samples.
Rechtliche Hinweise
Tube-O-Array is a trademark of Geno Technology, Inc.
Tube-O-DIALYZER is a trademark of Geno Technology, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)