Merck
Z740378
Eco-Friendly SpecTainer, tamper evident, biodegradable
size 90 mL, non-sterile
Größe auswählen
400 EA
CHF 533.00
CHF 533.00
Voraussichtliches Versanddatum28. Mai 2025
Größe auswählen
Ansicht ändern
400 EA
CHF 533.00
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.22
CHF 533.00
Voraussichtliches Versanddatum28. Mai 2025
Materialien
polyethylene cap
polypropylene container
Sterilität
non-sterile
Verpackung
pack of 400 ea
Hersteller/Markenname
Simport C566-90DOECO
Größe
90 mL
Allgemeine Beschreibung
The Simport Eco-Friendly SpecTainer will biodegrade to become some of the soil′s organic components in less than 7 years in constant contact with the soil instead of up to 400 years when using conventional plastics.
The Simport Eco-Friendly SpecTainer, one of the most innovative specimen containers on the market today, is now available in a new version: The Eco-Friendly SpecTainer, a rapidly degradable Specimen Container. Plastics take hundreds of years to degrade naturally in the environment. The only condition that is necessary for the Eco-Friendly SpecTainer to biodegrade is constant contact with other degrading material. No heat, physical stress, oxygen or sunlight necessary. The Eco-Friendly SpecTainer has the ability to break down safely and quickly, by biological means, into the raw materials of nature and disappear into the environment.
Does not contain heavy metals. No negative effect to its physical properties. No heat, light or mechanical stress needed for product breakdown. No toxic residue. Fully degradable within 7 years, aerobically or anaerobically.
Rechtliche Hinweise
SpecTainer is a trademark of Simport
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)