Merck
Z659290
Bandelin SONOPULS ultrasonic homogenizer accessories
Rosett cell RZ 2
Synonym(e):
Glass sample vessel
About This Item
UNSPSC-Code:
41101520
NACRES:
NB.74
Materialien
glass
Verpackung
pack of 1 ea
Hersteller/Markenname
Bandelin 3607
Verfügbarkeit
available only in EU
Volumen × ID × H
50 mL × 42 mm × 100 mm
Allgemeine Beschreibung
A rosette cell (RZ) enables intensive and uniform sonication of liquid media. The round shape of the side arms leads to an intensified circulation of the sample.
Leistungsmerkmale und Vorteile
  • Optimal fluid circulation .
  • Good heat dissipation .
  • Homogeneous sound reinforcement results
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)