Merck
Z514950
Kimax® high strength centrifuge tube replacement cap
for Z514896
Synonym(e):
73802-20400
Größe auswählen
1 PKG
CHF 250.00
CHF 250.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 PKG
CHF 250.00
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.11
CHF 250.00
Check Cart for Availability
Hersteller/Markenname
Kimble®
Leistungsmerkmale und Vorteile
  • Specially formulated phenolic resin and liner adhesive to withstand the effects of repeated autoclaving
  • PTFE faced/general purpose white rubber liners are highly resistant to chemical effects
  • Excellent for general laboratory use
  • Identified in ASTM Specification E982, Class A requirements
Rechtliche Hinweise
KIMBLE is a registered trademark of DWK Life Sciences
Kimax is a registered trademark of Gerresheimer Glass, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)