Merck
DWK73785-15
KIMBLE® centrifuge tube
with screw-top finish, conical bottom clear glass, capacity (15 mL), without cap, non-sterile
Größe auswählen
125 EA
CHF 222.00
CHF 222.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
125 EA
CHF 222.00
About This Item
UNSPSC-Code:
41121703
NACRES:
NB.11
CHF 222.00
Check Cart for Availability
Materialien
conical bottom clear glass
conical bottom
transparent borosilicate glass tube
Sterilität
non-sterile
Leistungsmerkmale
without cap
Verpackung
case of 125 ea Modular trays are shrink-wrapped
Hersteller/Markenname
Kimble® 73785-15
Kapazität
(15 mL)
Fitting
neck (External Thread)
Allgemeine Beschreibung
Screw Thread Disposable Centrifuge Tubes
  • These tubes are suitable for tissue culture work and general bacteriological use.
  • Supplied without caps
  • No marking spot
  • Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
Rechtliche Hinweise
KIMBLE is a registered trademark of DWK Life Sciences
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Questions
  1. matching cap for this tube?
    1 answer
    1. This product is manufactured by DWK Life Sciences, part number 73785-15 (found in the "Properties" section of the product page). According to the manufacturer, this product has a thread measurement of 13-415. The products that will fit this centrifuge tube are listed below:
      https://www.sigmaaldrich.com/product/aldrich/dwk45066c13
      https://www.sigmaaldrich.com/product/aldrich/dwk7380213415
      https://www.sigmaaldrich.com/product/aldrich/cls9999132
      https://www.sigmaaldrich.com/product/aldrich/cls9999913
      https://www.sigmaaldrich.com/product/aldrich/cls999813