Merck
SYNF224500
Synthware four neck round bottom flask with angled side necks
capacity 500 mL, center joint: ST/NS 24/40, side joint: ST/NS 24/40
Synonym(e):
Synthware F224500, 4-neck reaction flask
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.51
Materialien
borosilicate glass flask
four-neck
round bottom
Verbindung
center joint: ST/NS24/40
side joint: ST/NS24/40
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware F224500
Kapazität
500 mL
Allgemeine Beschreibung
Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)