Merck
SYND441410
Synthware Dean Stark distillation receiver with vacuum jacketed uptake tube
10 mL, joint: ST/NS 14/20
Synonym(e):
Synthware D441410,
Größe auswählen
1 EA
CHF 242.00
CHF 242.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 242.00
About This Item
UNSPSC-Code:
41104812
NACRES:
NB.51
CHF 242.00
Check Cart for Availability
Verbindung
joint: ST/NS14/20
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware D441410
Kapazität
10 mL
Allgemeine Beschreibung
Similar to D451414 above but with the uptake tube vacuum jacketed to improve distillation performance. The jacket eliminates the possibility of condensation or recrystallization of distillate in the sidearm. With 2 mm PTFE stopcock.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)