Merck
CLS5320250
Pyrex® heavy-wall graduated filtering flask
capacity 250 mL
Synonym(e):
conical flasks, filter flasks, filtering flask, filtering flasks, flask filter, Corning® 5320
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
Materialien
borosilicate glass
flat bottom flask
narrow-mouth
Verbindung
joint stopper6
Beschreibung
without hose connection
Sterilität
non-sterile
Leistungsmerkmale
closure type Rubber stopper
Verpackung
case of 18
pack of 6
Hersteller/Markenname
Corning 5320-250
Messbereich Volumen
75-250 mL
Kapazität
250 mL
Allgemeine Beschreibung
Blown in special molds to give maximum strength. Graduated to show approximate capacity.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)