Merck
C81004
Sigma-Aldrich
Zimtnitril
97%
Synonym(e):
3-Phenyl-acrylonitril
About This Item
Lineare Formel:
C6H5CH=CHCN
CAS-Nummer:
Molekulargewicht:
129.16
Beilstein:
1209546
EG-Nummer:
MDL-Nummer:
UNSPSC-Code:
12352100
PubChem Substanz-ID:
NACRES:
NA.22
Form:
liquid
Assay:
97%
Assay
97%
Form
liquid
Brechungsindex
n20/D 1.601 (lit.)
bp
254-255 °C (lit.)
mp (Schmelzpunkt)
18-20 °C (lit.)
Dichte
1.028 g/mL at 25 °C (lit.)
SMILES String
N#C\C=C\c1ccccc1
InChI
1S/C9H7N/c10-8-4-7-9-5-2-1-3-6-9/h1-7H/b7-4+
InChIKey
ZWKNLRXFUTWSOY-QPJJXVBHSA-N
Piktogramme
Skull and crossbones
Signalwort
Danger
H-Sätze
Gefahreneinstufungen
Acute Tox. 3 Oral
Lagerklassenschlüssel
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 2
Flammpunkt (°F)
235.4 °F - closed cup
Flammpunkt (°C)
113 °C - closed cup
Persönliche Schutzausrüstung
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Kunden haben sich ebenfalls angesehen
Slide 1 of 2
1 of 2
P J van Bladeren et al.
Drug metabolism and disposition: the biological fate of chemicals, 9(3), 246-249 (1981-05-01)
After administration of acrylonitrile, crotononitrile and cinnamonitrile to rats, two types of mercapturic acids were isolated from urine and identified by mass and NMR spectroscopy as N-acetyl-S-(2-cyanoethyl)-L-cysteine (I) and N-acetyl-S-(2-hydroxyethyl)-L-cysteine (II) (methyl-substituted in the case of crotonitrile and phenyl-substituted in
Deepali N Mehta-Hurt et al.
The Journal of chemical physics, 143(7), 074304-074304 (2015-08-25)
In Titan's atmosphere, photochemical pathways that lead to nitrogen heteroaromatics may incorporate photoisomerization of their structural isomers as a final step. (E)- and (Z)-phenylvinylnitrile ((E)- and (Z)-PVN, C6H5-CH=CHCN) are structural isomers of quinoline that themselves possess extensive absorptions in the
Yasushi Obora et al.
Organic & biomolecular chemistry, 8(18), 4071-4073 (2010-07-24)
A facile direct synthesis of cinnamonitriles from acrylonitriles and benzenes is successfully achieved by using Pd(OAc)(2)/HPMoV/O(2) catalyst system via the direct C-H bond activation of benzenes using molecular oxygen as a terminal oxidant.
Susanne Günther et al.
Journal of microbiological methods, 70(2), 272-283 (2007-06-15)
Cultivated bacterial toluene degraders use one or several of four described pathways for the aerobic degradation of this priority groundwater contaminant. To be able to identify un-cultivated toluene-degrading bacteria within enriched or natural consortia, we attempted to develop a set
Iana M Serafimova et al.
Nature chemical biology, 8(5), 471-476 (2012-04-03)
Targeting noncatalytic cysteine residues with irreversible acrylamide-based inhibitors is a powerful approach for enhancing pharmacological potency and selectivity. Nevertheless, concerns about off-target modification motivate the development of reversible cysteine-targeting strategies. Here we show that electron-deficient olefins, including acrylamides, can be