Merck
BR91214
BRAND® glass beaker with spout, low form
volume 25 mL, with graduation
Synonym(e):
Beakers, Glass
Größe auswählen
10 EA
CHF 93.20
CHF 93.20
Voraussichtliches Versanddatum30. Mai 2025
Größe auswählen
Ansicht ändern
10 EA
CHF 93.20
About This Item
UNSPSC-Code:
41121803
NACRES:
NB.43
CHF 93.20
Voraussichtliches Versanddatum30. Mai 2025
Materialien
borosilicate glass 3.3
Leistungsmerkmale
with graduation
Hersteller/Markenname
BRAND 91214
Volumen
25 mL
Allgemeine Beschreibung
The BRAND® Glass Beaker with Spout is expertly crafted from Boro 3.3 glass, making it a reliable choice for laboratory applications. Designed in accordance with DIN EN ISO 3819 standards, this low-form beaker features clear graduation markings for precise measurement of liquids. The integrated spout allows for easy pouring, minimizing spills and enhancing accuracy during liquid transfer.
Sonstige Hinweise
Browse all glass beakers
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)