Skip to Content
MilliporeSigma

Skip To

E2138

Echistatin

α1 isoform, ≥90% (HPLC)

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C217H341N71O74S9
CAS Number:
Molecular Weight:
5417.05
UNSPSC Code:
12352200
MDL number:
Form:
powder
Assay:
≥90% (HPLC)
Biological source:
Echis carinatus


biological source

Echis carinatus

assay

≥90% (HPLC)

form

powder

technique(s)

flow cytometry: suitable, inhibition assay: suitable

storage temp.

−20°C

General description

Echistatin is a single chain 49 amino acid residue protein, which prevents the aggregation of platelets. It has an isoelectric point (pI) of 8.3 and a molecular weight of 5400. This peptide is present in the venom of Echis carinatus, which is a saw-scaled viper. It contains the arginine-glycine-aspartic (RGD) acid sequence, which is present in proteins that bind to glycoprotein IIb/IIIa complex. It shares the proline-arginine-asparagine-proline sequence with the Aα chain of human fibrinogen.[1] This protein is a member of disintegrin family, which prevents cell adhesion.[2] With regards to molecular weight, echistatins are the smallest member of disintegrin family, and contains four isoforms called, α1, α2, β and γ.[3]

Application

Echistatin has been used:
  • as an inhibitor of integrin function to study the role of microfibril-associated glycoprotein-1 (Magp1) in the morphogenesis of vascular structures[4]
  • for the preparation of microbubbles targeted to αvβ3 integrins in tumor angiogenesis imaging[5]
  • as a conjugate to αvβ3 in flow cytometric binding studies[6]

Biochem/physiol Actions

A member of the disintegrins family, a novel family of integrin β1 and β3 inhibitor proteins and the most potent known inhibitors of integrin function.
Disintegrins represent a novel family of integrin β1 and β3 inhibitor proteins isolated from viper venoms. They are low molecular-weight, cysteine-rich peptides containing the Arg-Gly-Asp (RGD) sequence. They are the most potent known inhibitors of integrin function. Disintegrins interfere with cell adhesion to the extracellular matrix, including adhesion of melanoma cells and fibroblasts to fibronectin, and are potent inhibitors of platelet aggregation.
Echistatin is a disintegrin, which prevents the aggregation of platelets. They interact with and prevent the binding of fibrinogen to their receptors on the membrane of platelets.[3] It also inhibits platelet aggregation mediated by epinephrine, thrombin, collagen, or platelet-activating factor.[1] Studies in isolated osteoclasts show that this peptide inhibits bone resorption by osteoclasts, most probably by damaging adhesion structures.[7]


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Questions

Reviews

No rating value

Active Filters