MilliporeSigma
Mass Spectrometry
Mass spectrum (relative abundance vs. m/z ratio) of Man-3 glycan showing molecular ion peak, base peak, and fragmentation peaks
Mass spectrometry (MS) is an analytical technique that uses mass-to-charge (m/z) ratio to identify compounds in a sample. The method identifies a compound by determining its molecular weight and analyzing its isotopic abundance. A mass spectrometer ionizes the sample into gaseous ions and then identifies the ions by their mass-to-charge ratios and relative abundances.
Featured Categories
Chromatography & Spectroscopy Reagents
Chromatography & Spectroscopy Reagents
Explore our selection of high-quality analytical reagents for wet chemistry, chromatography, spectroscopy, analytical, titration, water quality control, and electrochemistry.
Shop Products
Thin Layer Chromatography (TLC)
Thin Layer Chromatography (TLC)
Choose from classical TLC plates with silica, aluminum, glass, and plastic backings as well as HPTLC plates, MS-Grade TLC and HPTLC plates, and PLC plates.
Shop Products
HPLC Columns
HPLC Columns
Achieve precise separations with our extensive HPLC column collection. Enhance retention, resolution, and selectivity. Order today.
Shop Products
Today, mass spectrometry is a well-established detection method that offers a multitude of benefits, such as selectivity, sensitivity, and multi-sample analysis. 
It can be coupled to various chromatographic techniques, such as liquid chromatographythin layer chromatographygas chromatography, or inductively coupled plasma. Mass spectrometry is widely used across many research fields and industries, including the pharmaceutical and food industries, health clinics, clinical research labs, and forensic and environmental testing labs.
How does mass spectrometry work?
A mass spectrometer works by converting individual molecules into ions and then analyzing the relative abundance of the generated ions. In the ion chamber of a mass spectrometer, each individual molecule is ionized to form a molecular ion, having one electron less than the parent molecule. Molecular ions, or ‘radical cations’, then undergo fragmentation into ions which, in turn, are further fragmented, and so on. From one complex sample, a mass spectrometer generates many ions. The ions are then accelerated in an electromagnetic field and separated based on their mass-to-charge (m/z) ratios. The instrument’s detector records the ions in proportion to their relative abundance and generates a mass spectrum of the molecule.
Applications of mass spectrometry
Due to the sensitivity of mass spectrometry, it is widely used to measure very low molecular weights at extremely low concentrations, below nanograms per milliliter (ng/mL). The ability to couple mass spectrometry to other separation techniques such as capillary electrophoresis, GC, and HPLC makes it a versatile analytical tool for the simultaneous separation and identification of analytes.
Typical applications of mass spectrometry include:
  • Analysis of amino acid sequences of proteins and peptides
  • Evaluation of impurities during drug development
  • Purity assessment of active pharmaceutical ingredients
  • Routine analysis of illegal drugs in urine, blood and hair
  • Detection of hereditary disorders of amino acid, fatty acid, and organic biosynthesis
Document Search
Looking for More Specific Information?
Visit our document search for data sheets, certificates and technical documentation.
Find Documents