Blood Plasma Fractionation
Plasma fractionators play a critical role in the treatment of rare life-threatening conditions. Consistent, reliable performance is key to plasma fractionation, posing challenges that must be met in order to better serve patients around the globe: improving yield, maintaining process economics, and managing ever-increasing regulatory requirements. Choosing the right partner with deep understanding of these challenges is vital, offering fractionators a gateway to new solutions and resources that can enhance their full range of blood plasma products.
Immunoglobulin
Polyclonal immunoglobulin G (IgG) is the primary high-value product for all plasma manufacturers. Today, immunoglobulin is administered in higher concentration dosages; this requires formulations which are well-tolerated by the patient. Key parameters for tolerability include:
- High purity
- Low immunoglobulin A and M (igA, IgM) content
- Low prekallikrein activator (PKA) content
- Low anticardiolipin antibody (ACA) content
- Low isoagglutinin content
The clinical efficacy of IgG is determined by the process and the quality of manufacture. Key features include an intact molecule, physiological sub-class distribution, pH of administration, absence of pyrogens, and low toxic residue content. IgG is currently used to treat primary immunodeficiency (PID), neurological conditions, hematology, and other infectious diseases.
Albumin
Albumin, often referred to as HSA (human serum albumen), is a stable protein with a molecular weight of 67 kD. It is sometimes added as a stabilizer for other plasma products, though its therapeutic use is as a fluid replacement treatment to expand blood volume in patients typically traumatized by burns or surgery. Albumin is manufactured in large quantities and is produced by all plasma manufacturers. Frequently, three to five batches per week may be manufactured in both low and high concentrations.
While raw plasma contains around 35–50 g of albumin per liter, 25–40% of that volume is lost during the actual fractionation process. This can lead fractionators to consider a chromatography method instead of Cohn fractionation, as chromatography has more favorable loss percentages of around 15–20%.
Factor VIII
Improving purity while maintaining specific activity is important in production of Factor VIII (FVIII), an essential blood-clotting protein. Clinical requirements of administration to a patient include low immunogenicity and high purity, which translates to low isoagglutinins and accompanying protein count, no foreign proteins or DNA, and low amounts of chemical residues. The purification process is designed to achieve these requirements:
- Increasing process efficiency and product recovery
- Achieving higher product yield
- Ensuring process safety and reproducibility of multi-strain products
Related Product Resources
- Application Note: Integration of Bioburden Reduction and Sterile Filtration in Human Plasma IgG Purification
- Brochure: Leveraging the Advantages of Single-use in Plasma Processing
- Brochure: Plasma Processing Capabilities
- Verification Report: Buffer Preparation for Plasma Processes: Process Efficiency
- White Paper: Intensification of Human Plasma IgG Purification for Intravenous and Subcutaneous Administration
- What is MilliporeSigma doing About REACH Compliance?
Explore our comprehensive portfolio of detergents and surfactants as possible Triton X-100 alternatives for your research applications before the REACH Annex 14 ban takes effect.
- Brochure: Process Development and Drug Manufacturing
We provide comprehensive services for drug development and manufacturing, including technical and regulatory expertise and process development support.