Skip to Content
MilliporeSigma

Skip To

D8143

2′-Deoxycytidylyl(3′→5′)-2′-deoxyguanosine ammonium salt

~95%

Synonym(s):

dCpdG

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C19H25N8O10P · xNH3
CAS Number:
Molecular Weight:
556.42 (free acid basis)
UNSPSC Code:
41106305
PubChem Substance ID:
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

~95%

form

solid

storage temp.

−20°C

SMILES string

N.NC1=NC(=O)c2ncn(C3CC(O)C(COP(O)(=O)OC4CC(OC4CO)N5C=CC(N)=NC5=O)O3)c2N1

InChI

1S/C19H25N8O10P.H3N/c20-12-1-2-26(19(31)23-12)14-4-9(10(5-28)35-14)37-38(32,33)34-6-11-8(29)3-13(36-11)27-7-22-15-16(27)24-18(21)25-17(15)30;/h1-2,7-11,13-14,28-29H,3-6H2,(H,32,33)(H2,20,23,31)(H3,21,24,25,30);1H3

InChI key

JZNMHBKGKVOTED-UHFFFAOYSA-N



pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library