Skip to Content
MilliporeSigma

Skip To

92011

Nitrile hydratase

recombinant, expressed in E. coli, ≥1.5 U/mg

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352204
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in E. coli

form

lyophilized

specific activity

≥1.5 U/mg

storage temp.

−20°C

Application

Nitrile hydratase is an enzyme that is used to convert nitriles to amides. It is used to study the metabolism of nitriles in bacteria, to produce acrylamide, and to remove nitriles from wastewater [1]. Product 92011 is used to convert acrylonitrile to acrylamide.

Biochem/physiol Actions

Nitrile hydratase catalyses the hydration of nitriles to their corresponding amides. Nitrile hydratase is composed of α- and β-subunits. It has a non-heme iron or a noncorrin cobalt ion in a ligand environment that includes two oxidized cysteine residues [1].

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

1 U corresponds to the amount of enzyme which converts 1 μmol acrylonitrile to acrylamide per minute at 30°C and pH 7.2


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Zhemin Zhou et al.
The Journal of biological chemistry, 284(22), 14930-14938 (2009-04-07)
The incorporation of cobalt into low molecular mass nitrile hydratase (L-NHase) of Rhodococcus rhodochrous J1 has been found to depend on the alpha-subunit exchange between cobalt-free L-NHase (apo-L-NHase lacking oxidized cysteine residues) and its cobalt-containing mediator (holo-NhlAE containing Cys-SO(2)(-) and



Global Trade Item Number

SKUGTIN
594202-25G04061832242590