Skip to Content
MilliporeSigma

Skip To

154792

Dichloromethane

ACS spectrophotometric grade, ≥99.5%, contains 50-150 ppm amylene as stabilizer

Synonym(s):

Methylene chloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
CH2Cl2
CAS Number:
Molecular Weight:
84.93
EC Number:
200-838-9
UNSPSC Code:
12352101
PubChem Substance ID:
Beilstein/REAXYS Number:
1730800
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

ACS spectrophotometric grade

vapor density

2.9 (vs air)

vapor pressure

24.45 psi ( 55 °C), 6.83 psi ( 20 °C)

assay

≥99.5%

form

liquid

autoignition temp.

1223 °F

contains

50-150 ppm amylene as stabilizer

expl. lim.

22 %

technique(s)

UV/Vis spectroscopy: suitable

impurities

≤0.0003 meq/g Titr. acid, ≤0.02% water

evapn. residue

≤0.002%

color

APHA: ≤10, clear

refractive index

n20/D 1.424 (lit.)

bp

39.8-40 °C (lit.)

mp

−95 °C (lit.)

density

1.325 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 235 nm Amax: 1.0, λ: 240 nm Amax: 0.35, λ: 250 nm Amax: 0.10, λ: 260 nm Amax: 0.04, λ: 340-400 nm Amax: 0.01

SMILES string

ClCCl

InChI

1S/CH2Cl2/c2-1-3/h1H2

InChI key

YMWUJEATGCHHMB-UHFFFAOYSA-N

Preparation Note

Product filtered through a 0.2 μm filter

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.


Still not finding the right product?

Explore all of our products under Dichloromethane


pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk

WGK 2

flash_point_f

does not flash

flash_point_c

does not flash

ppe

Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters