Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

EX0996

Supelco

300 Detergent phosphate-free

Extran®

Synonym(s):

Detergent

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$62.30
4 L
$184.00
20 L
$732.00

$62.30


Available to ship onApril 03, 2025Details


Request a Bulk Order

Select a Size

Change View
1 L
$62.30
4 L
$184.00
20 L
$732.00

About This Item

UNSPSC Code:
12161902

$62.30


Available to ship onApril 03, 2025Details


Request a Bulk Order

form

liquid

pH

11.5 ( in H2O, undiluted)

shipped in

ambient

storage temp.

room temp

Application

A powerful, liquid concentrate specially formulated to remove stubborn contamination and burnt-on residues. extran 300 is especially effective at removing organic materials such as blood and proteins. extran 300 is phosphate and halogen free and works well in ultrasonic cleaning baths. Can be used on equipment and apparatus made of glass, plastic, quartz, porcelain, ferrous metals and rubber. A final rinse with distilled water will leave a residue-free surface. Dose: 10 to 200mL per literpH of a 1% solution: 10.1

Legal Information

EXTRAN is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B P MacLeod et al.
Science advances, 6(20), eaaz8867-eaaz8867 (2020-05-20)
Discovering and optimizing commercially viable materials for clean energy applications typically takes more than a decade. Self-driving laboratories that iteratively design, execute, and learn from materials science experiments in a fully autonomous loop present an opportunity to accelerate this research

Questions

1–4 of 4 Questions  
  1. Is the room temperature storage requirement temperature 15-25°C? Or another range?

    1 answer
    1. Yes, 15-25°C is considered room temperature range.

      Helpful?

  2. Does Extran 300 contain any non-ionic detergents?

    1 answer
    1. Extran 300 detergent contains both: non-ionic and anionic detergents.

      Helpful?

  3. What is the pH of this cleaning detergent?

    1 answer
    1. The pH of the undiluted product is 11.5. When prepared at a rate of 10mL per 200mL distilled water, the pH is 10.1.

      Helpful?

  4. THE LAB SOAKS GLASS PIPETTES FOR 7 DAYS IN A DILUTE CLEANING SOLUTION. CAN THIS PRODUCT BE USED FOR THAT PURPOSE?

    1 answer
    1. This product is used to clean equipment and apparatus made of glass, plastic, quartz, porcelain, ferrous metals, and rubber. Glass pipettes can be soaked in this product to clean them. Extended exposure would not damage the pipettes. The time frame would need to be established by the end user.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service