Skip to Content
MilliporeSigma
All Photos(1)

Documents

EX0995

Supelco

MN01 Detergent Phosphate-free

powder, Extran®

Synonym(s):

cleaner

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103206

product line

Extran®

form

powder

manufacturer/tradename

Extran®

color

off-white

pH

7.0-9.0 (1% in water)

shipped in

ambient

storage temp.

room temp

General description

powder Extran®

Application

A multi-purpose powdered detergent for heavy-duty laboratory cleaning applications. extran® MN01 is a mixture of anionic surfactants and auxiliary agents. Can be used on glass, plastic, rubber, metal and quartz. Articles to be cleaned should be immersed in a cleaning solution and thoroughly brushed or wiped. Rinsing with distilled water will leave a residue-free surface.
powder Extran®

Analysis Note

AppearanceOff-white powderOdorMildpH (1%, water)7.0-9.0

Legal Information

EXTRAN is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service