MilliporeSigma
Phosphates
The image appears to be of a small white plastic bottle with a red cap, labeled “Phosphate buffered saline 10mL”.
The term phosphate can refer to a variety of anions, salts, esters, or functional groups derived from phosphoric acid (H3PO4), which can be formed by either the removal or substitution of one or more of phosphoric acid’s three acidic protons. Most phosphates exhibit solubility in water under standard temperature and pressure. The phosphates of sodium, potassium, rubidium, cesium, and ammonium are all soluble in water, while others are either insoluble or only sparingly soluble. Hydrogen and dihydrogen phosphates generally exhibit higher solubility compared to their corresponding phosphates.
Phosphates play a significant role in biological systems as the essential components of teeth and bones. Beyond their structural significance, phosphates serve multifaceted roles in biological systems, acting as crucial building blocks for DNA, RNA, and cellular structures like phospholipids, ensuring overall structural integrity. Additionally, phosphates function as buffers, contributing to stable pH levels in cell culture media and creating optimal conditions for cell growth and viability.
Read more about: 
Phosphates-as-Building-Blocks
Phosphates as Buffer
Calcium Phosphate Microcarriers in Bone Tissue Engineering
Products
Sort by Relevance
Showing 1-20 of 1102
Change View
Showing 1-20 of 1102
Compare
Product Number
Product Name
Product Description
Pricing
Phosphates as Building Blocks
Organic building blocks consist of functionalized organic molecules, serving as the fundamental components for organic synthesis. Phosphates play a pivotal role as building blocks in DNA and RNA, responsible for carrying genetic information in all living organisms. They can serve as leaving groups in substitution reactions, and their diverse reactivity makes them valuable intermediates in the construction of complex organic molecules.
Phosphates as Buffer
Phosphate buffer is a liquid medium designed for the qualitative detection of microorganisms in pharmaceutical and food samples. It serves as a sample diluent and as a medium for performing serial dilution. It prevents pH-associated variations while carrying out microbial experiments.
Potassium phosphates 
Potassium phosphate buffers are widely employed in equilibrium dialysis, high-performance liquid chromatography (HPLC), and affinity capillary electrophoresis studies.
Potassium phosphate monobasic is a water-soluble inorganic salt widely employed in various applications, contributing significantly to the formulation of phosphate buffer and phosphate-buffered saline (PBS). It has also proven valuable in the preparation of PEG1000 (polyethylene glycol)/potassium phosphate aqueous two-phase systems (ATPS).
Potassium phosphate dibasic is a useful biological buffer having a pH in the range of 8.5–9.6. It has been used in the preparation of MMG (maltose, methyl green) medium, phosphate buffered saline, and hypotonic solution.
Sodium phosphates 
Sodium phosphates are used as thickening agents, leavening agents, and emulsifiers in processed cheese. They are also utilized in medicine for constipation treatment and bowel preparation before surgeries. They are effective anti-rust agents and are incorporated into detergents to soften water. Sodium phosphate is a widely used buffer in molecular biology, biochemistry, and chromatographic studies.
Sodium phosphate monobasic can also be used as a catalyst in the synthesis of non-ionic surfactants for adhesive and detergent applications. Additionally, it is used as a mobile phase in High-performance liquid chromatography (HPLC) for various analyses, including the determination of adenosine in human synovial fluid, analysis of neuroendocrine tumor markers by HPLC-electrochemical detection, and the analysis of benzalkonium chloride and its homologs using HPLC/HPCE.
Calcium Phosphate Microcarriers in Bone Tissue Engineering
Microcarriers (MCs) are commonly used in tissue engineering to provide cells with a significant surface area, facilitating the high-yield culture of anchorage-dependent cells. Calcium phosphates, composed of calcium cations and phosphate anions, constitute approximately 60% of the inorganic composition of all-natural human bones. These minerals significantly impact bioactivity in osteoblasts, affecting processes such as adhesion, proliferation, and new bone production.
Calcium phosphates also play important roles in cell adhesion and tissue formation by affecting the adsorption of extracellular matrix proteins on the surface. The characteristics of calcium phosphates impact the mineralization of newly formed bone, consequently influencing bone regeneration.