Skip to Content
MilliporeSigma
All Photos(1)

Documents

Z742916

Liquinox® phosphate-free liquid detergent

3.8 L packs

Synonym(s):

USDA authorized detergent, anionic detergent, biodegradable cleaner, free rinsing cleaner, ultrasonic detergent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131800
NACRES:
NB.34

packaging

pkg of 3.8 L
pkg of 4 × 3.8 L

manufacturer/tradename

Alconox, Inc. 1201 (4 x 3.8 L)
Alconox, Inc. 1201-1 (3.8 L)

technique(s)

tissue culture: suitable

application(s)

detection

Looking for similar products? Visit Product Comparison Guide

General description

Concentrated, anionic detergent for manual and ultrasonic cleaning. Free rinsing to give you reliable results without interfering residues. Extremely mild and completely soluble in hard and soft water. USDA authorized. Dilute 1:100. pH 8.5

Legal Information

Liquinox is a registered trademark of Alconox, Inc.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service