Skip to Content
MilliporeSigma
All Photos(3)

Documents

1.07555

Supelco

Extran® MA 01

greener alternative

liquid, alkaline, concentrate

Synonym(s):

TM=["EXTRAN"] MA 01 cleaning agent; TM=["EXTRAN"] MA 01 detergent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103206
NACRES:
NA.21

vapor pressure

<23.4 hPa ( 20 °C)

Quality Level

form

liquid

greener alternative product characteristics

Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

pH

12.94 (20 °C in H2O, undiluted)

bp

>100 °C/1013 hPa

density

1.07 g/cm3 at 20 °C

greener alternative category

storage temp.

no temp limit

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is environmentally friendly having bio-degradable active ingredients. Click here for more information.

Linkage

Replaces: EX0993; EX0993-4

Analysis Note

pH-value (5 %; water): 11.0 - 12.0
Density (d 20 °C/ 4 °C): 1.065 - 1.075
Foaming volume (5 %; water): ≥ 70 ml

Legal Information

EXTRAN is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service