Skip to Content
MilliporeSigma
All Photos(2)

Documents

Z742914

Alconox® detergent

1.8Kg cartons

Synonym(s):

anionic detergent, concentrated powdered detergent, general laboratory cleaner, lab detergent, low foaming detergent, ultrasonic cleaner detergent

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
47131800
NACRES:
NB.34

packaging

pkg of 1.8 kg
pkg of 9 × 1.8 kg

manufacturer/tradename

Alconox, Inc. 1104-1 (1.8 KG)
Alconox, Inc. 1104 (9 x 1.8 KG)

Looking for similar products? Visit Product Comparison Guide

General description

ALCONOX® is a mixture of a homogeneous blend of sodium linear alkylaryl sulfonate, alcohol sulfate, phosphates, and carbonates.
Powdered detergent for general lab cleaning.

Features and Benefits

  • Concentrated and long-lasting
  • Biodegradable and readily disposable
  • Penetrating wetting power to save
  • Alternative for corrosive acids and hazardous solvents
  • Easy to rinse, doesn′t form interfering residues, and provides reliable results

Legal Information

ALCONOX is a registered trademark of Alconox, Inc.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service