Skip to Content
MilliporeSigma

Skip To

8.02458

4-Chlorostyrene

(stabilised with 4-tert-butylpyrocatechol) for synthesis

Synonym(s):

4-Chlorostyrene, 1-Chloro-4-ethenyl-bezene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 mL
Please contact Customer Service for Availability
$105.00

About This Item

Empirical Formula (Hill Notation):
C8H7Cl
CAS Number:
Molecular Weight:
138.59
UNSPSC Code:
12352101
EC Index Number:
214-028-8
NACRES:
NA.22
MDL number:

$105.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98.0% (GC)

form

liquid

potency

5200 mg/kg LD50, oral (Rat)

bp

192 °C

transition temp

flash point 68 °C

density

1.09 g/cm3 at 20 °C

storage temp.

2-8°C

SMILES string

Clc1ccc(cc1)C=C

InChI

1S/C8H7Cl/c1-2-7-3-5-8(9)6-4-7/h2-6H,1H2

InChI key

KTZVZZJJVJQZHV-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.085 - 1.090
Identity (IR): passes test

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.02254651990111961
assay

≥98.0% (GC)

assay

≥98.0% (GC)

assay

97%

assay

95%

storage temp.

2-8°C

storage temp.

2-30°C

storage temp.

-

storage temp.

-

density

1.09 g/cm3 at 20 °C

density

1.26 g/cm3 at 45 °C

density

1.5080 g/mL at 25 °C (lit.)

density

-

form

liquid

form

crystals

form

-

form

solid

bp

192 °C

bp

220 °C/1013 hPa

bp

76-78 °C/0.3 mmHg (lit.)

bp

216-222 °C (lit.)

potency

5200 mg/kg LD50, oral (Rat)

potency

-

potency

-

potency

-


Still not finding the right product?

Explore all of our products under 4-Chlorostyrene


pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Aquatic Chronic 2 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

154.4 °F - closed cup

flash_point_c

68 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters