Merck
  • The effect of steric changes on the isoselectivity of dinuclear indium catalysts for lactide polymerization.
The effect of steric changes on the isoselectivity of dinuclear indium catalysts for lactide polymerization.
Dalton transactions (Cambridge, England : 2003) (2015-03-03)
K M Osten, D C Aluthge, P Mehrkhodavandi
RESUMO
A series of (±)- and (R,R)-tridentate diamino, ortho/para disubstituted phenolate proligands H(NNO(R)) with various phenolate substituents was synthesized and used to prepare indium dichloride complexes (NNO(R))InCl2via salt metathesis of the deprotonated ligands with indium trichloride. These complexes are dinuclear in the solid state, in contrast to previously reported complexes with t-butyl or methyl phenolate substituents. Solution state (1)H and PGSE NMR spectroscopy suggests that a fast exchange between the monomeric and dimeric forms of these complexes may exist in solution and is likely influenced by the chirality of the complexes undergoing aggregation. The indium dichloride complexes were utilized to synthesize dinuclear indium ethoxide complexes via salt metathesis with sodium ethoxide. These complexes were active for the polymerization of lactide. In situ and bulk polymerization data confirmed differences in the activity and selectivity of these systems based on the phenolate substituents as well as the ligand chirality.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Diclorometano, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Diclorometano, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Diclorometano, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Diclorometano, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Sigma-Aldrich
Diclorometano, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Diclorometano, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
Sigma-Aldrich
Diclorometano, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Diclorometano, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Supelco
Diclorometano, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Diclorometano, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Deuterium, 99.9 atom % D
Supelco
Diclorometano, analytical standard
Supelco
Diclorometano, Selectophore, ≥99.5%
Sigma-Aldrich
Diclorometano, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer