Merck
  • Development of andrographolide loaded PLGA microspheres: optimization, characterization and in vitro-in vivo correlation.
Development of andrographolide loaded PLGA microspheres: optimization, characterization and in vitro-in vivo correlation.
International journal of pharmaceutics (2014-09-16)
Yunxia Jiang, Fang Wang, Hui Xu, Hui Liu, Qingguo Meng, Wanhui Liu
RESUMO
The purpose of this study was to develop a sustained-release drug delivery system based on the injectable PLGA microspheres loaded with andrographolide. The andrographolide loaded PLGA microspheres were prepared by emulsion solvent evaporation method with optimization of formulation using response surface methodology (RSM). Physicochemical characterization, in vitro release behavior and in vivo pharmacokinetics of the optimized formulation were then evaluated. The percent absorbed in vivo was determined by deconvolution using the Loo-Riegelman method, and then the in vitro-in vivo correlation (IVIVC) was established. Results showed that the microspheres were spherical with a smooth surface. Average particle size, entrapment efficiency and drug loading were found to be 53.18±2.11 μm, 75.79±3.02% and 47.06±2.18%, respectively. In vitro release study showed a low initial burst release followed by a prolonged release up to 9 days and the release kinetics followed the Korsmeyer-Peppas model. After a single intramuscular injection, the microspheres maintained relatively high plasma concentration of andrographolide over one week. A good linear relationship was observed between the in vitro and in vivo release behavior (R(2)=0.9951). These results suggest the PLGA microspheres could be developed as a potential delivery system for andrographolide with high drug loading capacity and sustained drug release.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Diclorometano, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Diclorometano, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Diclorometano, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Diclorometano, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Sigma-Aldrich
Diclorometano, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Diclorometano, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
Sigma-Aldrich
3,6-Dimethyl-1,4-dioxane-2,5-dione, 99%
Sigma-Aldrich
Diclorometano, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Diclorometano, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Glycolide, ≥99%
Supelco
Diclorometano, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Diclorometano, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Diclorometano, analytical standard
Supelco
Diclorometano, Selectophore, ≥99.5%
Sigma-Aldrich
Diclorometano, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer