Merck
  • Ecological and human health hazards of heavy metals and polycyclic aromatic hydrocarbons (PAHs) in road dust of Isfahan metropolis, Iran.
Ecological and human health hazards of heavy metals and polycyclic aromatic hydrocarbons (PAHs) in road dust of Isfahan metropolis, Iran.
The Science of the total environment (2014-12-03)
Naghmeh Soltani, Behnam Keshavarzi, Farid Moore, Tahereh Tavakol, Ahmad Reza Lahijanzadeh, Nemat Jaafarzadeh, Maryam Kermani
RESUMO
This study investigates trace elements and PAHs content in road dust of Isfahan metropolis, central Iran. The mean concentrations of As, Cd, Cu, Ni, Pb, Sb and Zn are 22.15, 2.14, 182.26, 66.63, 393.33, 6.95 and 707.19 mg kg(-1), respectively. When compared with upper continental crust, the samples generally display elevated trace element concentrations, except for Co and Cr. The decreasing trend of calculated enrichment factors (EFs) is Cd>Pb>Sb>Zn>Cu>As>Ni>Cr>Co. Calculated potential ecological risk reveals that among the analyzed metals, Cd and Pb, have a higher potential ecological risk. Statistically, two identified main sources of trace elements include road traffic emissions and resuspension of soil particles. As, Cd, Cu, Pb, Sb and Zn in Isfahan road dust are strongly influenced by anthropogenic activity, mainly traffic emissions, while Co, Cr and Ni originate from resuspension of soil natural parent particles. The sum of 13 major PAHs (∑13PAHs) mass concentration ranges from 184.64 to 3221.72 μg kg(-1) with the mean being 1074.58 μg kg(-1). PAHs sources are identified using PCA analysis. It is demonstrated that the PAHs in Isfahan road dust are mainly derived from traffic emission, coal combustion and petroleum. Toxic equivalent concentrations (TEQs) of PAHs in the road dust ranges between 25.021 μg kg(-1) and 230.893 μg kg(-1). High correlation coefficients (r(2)=0.909 and 0.822, p<0.01) between Benzo[a]pyrene, Benzo[b+k]fluoranthene and toxicity equivalent concentrations of road dust indicate that Benzo[a]pyrene and Benzo[b+k]fluoranthenes are major TEQ contributors. The total incremental life time cancer risk (ILCR) of exposure to PAHs from Isfahan metropolis urban dust is 4.85 × 10(-4) for adult and 5.02 × 10(-4) for children. Estimated results of ILCR indicate that Isfahan residents are potentially exposed to high cancer risk via both dust ingestion and dermal contact.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Diclorometano, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Diclorometano, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Diclorometano, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Hexano, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
Hexano, suitable for HPLC, ≥95%
Sigma-Aldrich
Hexano, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexano, Laboratory Reagent, ≥95%
Sigma-Aldrich
Hexano, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Hexano, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Diclorometano, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Sigma-Aldrich
Diclorometano, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Phenanthrene, 98%
Sigma-Aldrich
Diclorometano, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
Sigma-Aldrich
Diclorometano, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Acenaphthene, 99%
Sigma-Aldrich
Diclorometano, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Supelco
Diclorometano, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Diclorometano, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Hexano, anhydrous, 95%
Supelco
Hexano, analytical standard
Supelco
Phenanthrene, analytical standard, for environmental analysis
Sigma-Aldrich
Phenanthrene, sublimed grade, ≥99.5%
Supelco
Diclorometano, analytical standard
Supelco
Acenaphthene, analytical standard
Supelco
Diclorometano, Selectophore, ≥99.5%
Sigma-Aldrich
Diclorometano, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Phenanthrene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Acenaphthene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland