Merck
Role of combined zinc, vitamin A, and fish oil supplementation in childhood tuberculosis.
The Southeast Asian journal of tropical medicine and public health (2014-01-21)
Vollico Nenni, Heda Melinda Nataprawira, Tetty Yuniati
RESUMO
This objective of this study was to determine benefit of one month combined supplementation (zinc, vitamin A, fish oil) along with anti-tuberculosis drugs (ATD) on increasing serum leptin levels and decreasing tumor necrosis factor-alpha (TNF-alpha) in children with tuberculosis (TB). A quasi experimental study was conducted on 22 children (aged 5-14 years) with a positive acid-fast bacilli (AFB) smear. The children were divided into 2 groups. A history, physical examination, anthropometric measurements, serum leptin levels, TNF-alpha levels, retinol and zinc levels were examined in all subjects before and after treatment. Nutritional supplementation and ATD were given to group I while ATD only were given to group II. The change in leptin, TNF-alpha, retinol and zinc levels were analyzed with the Mann-Whitney test, while a t-test was used to determine changes in body mass index (BMI). Group I had a higher significant increase in serum leptin levels than group II (p=0.034). Group I had a significantly greater decrease in TNF-a levels than group II (p=0.032). No significant differences in retinol or zinc levels were seen between the two, but both groups had an increase after treatment. Both groups had a significant increase in BMI (p=<0.001) post-treatment compared to pre-treatment. Supplementation with zinc, vitamin A and fish oil is associated with a significant increase in leptin levels and a significant decrease in TNF-alpha levels among children treated for TB. No significant benefit was seen in BMI among children receiving supplementation compared to those without it, although ATD resulted in a significant increase in BMI in both groups.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Retinol, synthetic, ≥95% (HPLC), (Powder or Powder with Lumps)
Sigma-Aldrich
Zinc, dust, <10 μm, ≥98%
Sigma-Aldrich
Retinol, BioXtra, ≥97.5% (HPLC), ~3100 U/mg
Sigma-Aldrich
Zinc, powder, <150 μm, 99.995% trace metals basis
Sigma-Aldrich
Retinol, ≥95.0% (HPLC), ~2700 U/mg
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Zinc, granular, 30-100 mesh, 99%
Sigma-Aldrich
Zinc, granular, 20-30 mesh, ACS reagent, ≥99.8%
Sigma-Aldrich
Zinc, mossy, ≥99%
Sigma-Aldrich
Zinc preparation, 5 g/dL Zn+2 in THF, highly reactive Rieke®metal
Sigma-Aldrich
Zinc, sticks, diam. 7-10 mm, 99.97% trace metals basis
Sigma-Aldrich
Zinc, shot, <12 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, shot, 5 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Sigma-Aldrich
Zinc, purum, powder
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular