Merck
Examining the risk of methanol poisoning from methyl acetate-containing products.
The American journal of emergency medicine (2013-04-23)
Alicia B Minns, Anesia McIlvoy, Alycia Clark, Richard F Clark, F Lee Cantrell
RESUMO
This retrospective study aims to review California Poison Control System data to qualitatively describe reported methyl acetate (MA) exposures and determine if a metabolic acidosis develops. We queried the Poison Control System data between January 1997 and December 1, 2010. Inclusion criteria were single MA ingestions. Eighty-three cases were analyzed. Females made up 52% of study subjects. Seventy-five cases (90%) were 5 years or younger. Most (93%) ingestions were unintentional. Sixty-two cases (75%) were referred to a health care facility (HCF). Of these, 75% of cases referred to an HCF had no effect, and 25% of cases had a minor effect. There were no major effects, including deaths reported. One case received single-dose activated charcoal, and 1 case received intravenous fluids as therapy. All other cases were observed only. Of all cases, including home-managed and HCF cases, 63 (76%) had no symptoms. Vomiting was observed in 12 cases (14%), drowsiness in 1 case, ataxia in 1 case, abdominal pain in 1 case, and throat/oral irritation in 5 cases. Fifty-three percent of cases referred to HCF had at least 1 chemistry panel done (27% had >1 chemistry panel done). One patient had a mild metabolic acidosis without symptoms that resolved after a period of observation. There was no renal dysfunction. These data suggest that minor ingestions of MA may be observed at home, but a prospective study is needed to exclude the risk of home observation.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Metanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Metanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Metanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Metanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Metanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Metanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Metanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Metanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Metanol, BioReagent, ≥99.93%
Sigma-Aldrich
Metanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methyl acetate, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Metanol, Absolute - Acetone free
Sigma-Aldrich
Methyl acetate, ReagentPlus®, 99%
Sigma-Aldrich
Metanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Metanol, anhydrous, 99.8%
Sigma-Aldrich
Methyl acetate, ≥98%, FG
Sigma-Aldrich
Metanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Supelco
Metanol, analytical standard
Supelco
Methyl acetate, analytical standard
Sigma-Aldrich
Metanol, NMR reference standard
Sigma-Aldrich
Methyl acetate, anhydrous, 99.5%
Sigma-Aldrich
Methyl acetate, natural, 98%, FG
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.