Documentos Principais
Z329738
BRAND® wash bottle, wide-mouth, labelled, LDPE
capacity 250 mL, label, Distilled water, without venting valve
Selecione um tamanho
5 EA
R$ 411,00
R$ 411,00
Check Cart for Availability
Selecione um tamanho
Alterar visualização
5 EA
R$ 411,00
About This Item
Código UNSPSC:
41103209
NACRES:
NB.17
R$ 411,00
Check Cart for Availability
Materiais
low-density polyethylene bottle
natural bottle
round bottle
white closure
wide-mouth bottle
Características
label Distilled water
with cap
without venting valve
embalagem
pack of 5 ea
fabricante/nome comercial
BRAND 144078
capacidade
250 mL
Descrição geral
Flexible, high-quality wash bottles with integral wash steam and sealing cone in the closure. Large opening for convenient filling. Label with designation of contents and general safety statements (with CAS no. and NFPA codes).
Bottles with venting valve (metal ball) are for distilled water and organic solvents only; to avoid dropping caused by overpressure.
Outras notas
BR144078 (India)
Informações legais
BRAND is a registered trademark of BRAND GMBH + CO KG
acessório
Nº do produto
Descrição
Preços
Escolha uma das versões mais recentes:
Certificados de análise (COA)