Documentos Principais
Z278416
Sigma-Aldrich
Heat transfer block
Three bowl-shaped recesses
Selecione um tamanho
1 EA
R$ 444,00
R$ 444,00
Check Cart for Availability
Selecione um tamanho
Alterar visualização
1 EA
R$ 444,00
About This Item
Código UNSPSC:
41102404
NACRES:
NB.43
R$ 444,00
Check Cart for Availability
Características
Holds 3 x flasks
fabricante/nome comercial
Ace Glass 9594-16
Ace Glass 959416
Descrição geral
Aluminum blocks have excellent heat transfer rates and will not interfere with magnetic stirring. Replaces glass sand baths in microscale experiments. Use on top of stirring/hot plates.
Will fit 3 and 5 mL thin-walled vials and 5, 10, and 25 mL round-bottom flasks. Three bowl shaped recesses, one 20 mm straight-through hole, one deep bowl-shaped hole, and one hole for a thermometer. 31/2 in. square × 3/4 in. thick.
Escolha uma das versões mais recentes:
Certificados de análise (COA)