Documentos
Z108332
Leather gloves
size M
About This Item
Código UNSPSC:
46181504
NACRES:
NB.35
Materiais
brown gloves
leather
tamanho
M
esterilidade
non-sterile
Características
latex free
powder-free
reusable
embalagem
pack of 1 pair
Descrição geral
Constructed of specially treated leather that stays supple even at temperature extremes of -70 to 150°C. Remarkably durable, moisture, heat, and flame resistant. Provided with an elastic cinch for a no-slip fit.