Merck
R662461
4-AMINO-2-(2-OXO-2H-CHROMEN-3-YL)-5H-CHROMENO(4,3-D)PYRIMIDIN-5-ONE
AldrichCPR
About This Item
Fórmula linear:
C20H11N3O4
Número CAS:
Peso molecular:
357.328
Número MDL:
Preço e disponibilidade não estão disponíveis no momento.
Comparar Itens Semelhantes
Ver Comparação Total
Mostrar diferenças
1 of 4
Este Item
HPA052973HPA059419HPA044329
Quality Level
100
Quality Level
100
Quality Level
-
Quality Level
100
biological source
rabbit
biological source
rabbit
biological source
rabbit
biological source
rabbit
antibody form
affinity isolated antibody
antibody form
affinity isolated antibody
antibody form
affinity isolated antibody
antibody form
affinity isolated antibody
product line
Prestige Antibodies® Powered by Atlas Antibodies
product line
Prestige Antibodies® Powered by Atlas Antibodies
product line
Prestige Antibodies® Powered by Atlas Antibodies
product line
Prestige Antibodies® Powered by Atlas Antibodies
UniProt accession no.
Q8NDZ0
UniProt accession no.
Q5SZJ8
UniProt accession no.
Q6ZN30
UniProt accession no.
Q92805
Outras notas
Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of rare and unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH'S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT AS-IS AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
Escolha uma das versões mais recentes:
Certificados de análise (COA)