Documentos Principais
CAD7932
Laboratory pipetting needle with 90° blunt ends
gauge 20, L 1 1/2 in., nickel plated hub
Selecione um tamanho
12 EA
R$ 2.431,00
R$ 2.431,00
Check Cart for Availability
Selecione um tamanho
Alterar visualização
12 EA
R$ 2.431,00
About This Item
Código UNSPSC:
41122002
NACRES:
NB.03
R$ 2.431,00
Check Cart for Availability
Materiais
nickel plated hub
embalagem
pack of 12 ea
fabricante/nome comercial
Cadence Science 7932
Comprimento
1 1/2 in.
medidor
20
Categorias relacionadas
Descrição geral
Pipetting needles are available in a wide choice of sizes for use in laboratory procedures. All needles are made with type 304 stainless steel cannula.
Catalog numbers CAD7950 to CAD7957 needles are fitted with stainless steel hubs for special applications with corrosive materials or critical procedures. Catalog numbers CAD7930 to CAD7942 needles are fitted with standard nickel plated Luer hubs. These needles are used in conjunction with both manual and automatic volumetric dispensers, dilutors, and pipettors.
Escolha uma das versões mais recentes:
Certificados de análise (COA)