Documentos
BR129748
BRAND® PE-LD Media Bottle with screw cap
capacity 250 mL
About This Item
Código UNSPSC:
41121800
NACRES:
NB.13
Materiais
low-density polyethylene bottle
round bottle
Junta
Joints threaded neck (GL40)
Características
graduations: no
with cap
embalagem
pack of 1 ea
fabricante/nome comercial
BRAND 129748
capacidade
250 mL
Informações legais
BRAND is a registered trademark of BRAND GMBH + CO KG