Principaux documents
BR470816
BRAND® cover glass
L × W 24 mm × 40 mm, rectangular
Synonyme(s) :
Cover glasses, Microscopy
1000 EA
94,60 €
94,60 €
Disponibilité
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1000 EA
94,60 €
About This Item
Code UNSPSC :
41122600
Nomenclature NACRES :
NB.16
94,60 €
Disponibilité
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
borosilicate glass
rectangular
Fabricant/nom de marque
BRAND 470816
L × l
24 mm × 40 mm
Épaisseur
0.13-0.17 mm , No. 1
Description générale
Clear borosilicate glass, hydrolytic class 1, with excellent chemical resistance. Thickness No. 1 (0.13 to 0.17 mm). Refractive index ne = 1.52 ± 0.01, Abbe number ve = 56.5 ± 0.5. Full automatic production guarantees "ready-for-use" quality - clean, dust and grease free, distortion-free, flatness within ± 3 µm.
Caractéristiques et avantages
  • BRAND® borosilicate cover glass offers superior clarity and excellent chemical resistance.
  • Cover glasses for microscope slides are clean, dust/grease free and distortion-free.
  • Full automatic production guarantees "ready-for-use" quality.
  • Flatness within ± 3 μm.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG