Documents
BR462500
BRAND® rubber conical gasket for filter funnels and filter flasks
size 22-84 mm
Synonyme(s) :
Filter, Rubber gasket
About This Item
Code UNSPSC :
41104900
Nomenclature NACRES :
NB.24
Matériaux
EPDM rubber
Conditionnement
pack of 8 ea
Fabricant/nom de marque
BRAND 462500
Taille
22-84 mm
Description générale
Conical gaskets to place between filter funnel and filter flask.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG